missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBF-1 Polyclonal Antibody

Description
Immunogen sequence: WKLLSQKEKD AYHKKCDQKK KDYEVELLRF LESLPEEEQQ RVLGEEKMLN INKKQATSPA SKKPAQEGGK GGSEKPKRPV SAMFIFSEEK RRQLQEERPE LSESELTRLL ARMWNDLSEK KK Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
UBF-1 encodes a member of the HMG-box DNA-binding protein family. The encoded protein plays a critical role in ribosomal RNA transcription as a key component of the pre-initiation complex, mediating the recruitment of RNA polymerase I to rDNA promoter regions. The encoded protein may also play important roles in chromatin remodeling and pre-rRNA processing, and its activity is regulated by both phosphorylation and acetylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3, 11 and X and the long arm of chromosome 11.
Specifications
Specifications
| Antigen | UBF-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UBTF |
| Gene Accession No. | P17480, P25976 |
| Gene Alias | 90-kDa nucleolus organizer region autoantigen; A930005G04Rik; Autoantigen NOR-90; NOR90; NOR-90; Nucleolar transcription factor 1; Tcfubf; transcription factor UBF; UBF; UBF1; UBF-1; UBF2; Ubtf; upstream binding transcription factor, RNA polymerase I; upstream-binding factor 1 |
| Gene Symbols | UBTF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?