missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE4B Recombinant Rabbit Monoclonal Antibody (24GB1765)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen UBE4B
Clone 24GB1765
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275817
GREENER_CHOICE
View Documents
Invitrogen™
MA557824
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274842
View Documents
Invitrogen™
MA557825
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

UBE4B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications

Specifications

UBE4B
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
UBE4B
4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p
UBE4B
Primary
Affinity Chromatography
24GB1765
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95155, Q9ES00
10277, 298652, 63958
A synthesized peptide derived from human UBE4B (aa 15-50).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.