missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-58802
This item is not returnable.
View return policy
Description
UBE4B Polyclonal specifically detects UBE4B in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| UBE4B | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| AAD02233.1 | |
| UBE4B | |
| The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human ubiquitination factor E4B using the numbering given in entry AAD02233.1 (GeneID 10277). | |
| 0.1 mL | |
| Apoptosis | |
| 10277 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction