missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35766-20ul
This item is not returnable.
View return policy
Description
UBE4B Polyclonal antibody specifically detects UBE4B in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| UBE4B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-250 of human UBE4B (NP_006039.2).,, Sequence:, EVDENDRREKRSLSDKEPSSGPEVSEEQALQLVCKIFRVSWKDRDRDVIFLSSLSAQFKQNPKEVFSDFKDLIGQILMEVLMMSTQTRDENPFASLTATSQPIAAAARSPDRNLLLNTGSN | |
| 20 μL | |
| Apoptosis | |
| 10277 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction