missing translation for 'onlineSavingsMsg'
Learn More

UBE4A Antibody (1G8), Novus Biologicals™

Product Code. 18327588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327588 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327588 Supplier Novus Biologicals Supplier No. H00009354M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE4A Monoclonal antibody specifically detects UBE4A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE4A
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1G8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004779
Gene Alias E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A
Host Species Mouse
Immunogen UBE4A (NP_004779, 974 a.a. ~ 1073 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LAERIKSLADLQQQEEETYADACDEFLDPIMSTLMCDPVVLPSSRVTVDRSTIARHLLSDQTDPFNRSPLTMDQIRPNTELKEKIQRWLAERKQQKEQLE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9354
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.