missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE3C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531350
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (88%), Chicken (87%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 ubiquitin-protein ligase that accepts ubiquitin from the E2 ubiquitin-conjugating enzyme UBE2D1 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Can assemble unanchored poly-ubiquitin chains in either 'Lys-29'- or 'Lys-48'-linked polyubiquitin chains. Has preference for 'Lys-48' linkages. It can target itself for ubiquitination in vitro and may promote its own degradation in vivo.
Specifications
| UBE3C | |
| Polyclonal | |
| Unconjugated | |
| UBE3C | |
| AI853514; HectH2; HECT-type ubiquitin transferase E3C; KIAA0010; KIAA10; mKIAA0010; RGD1559986; Ube3c; ubiquitin protein ligase E3C; ubiquitin-protein isopeptide ligase (E3); ubiquitin-protein ligase E3C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100763, 9690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15386, Q80U95 | |
| UBE3C | |
| Recombinant protein encompassing a sequence within the center region of human UBE3C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction