missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UBE3C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29419
This item is not returnable.
View return policy
Description
UBE3C Polyclonal antibody specifically detects UBE3C in Human samples. It is validated for Immunoprecipitation
Specifications
| UBE3C | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 6.3.2.-, KIAA0010KIAA10ubiquitin-protein isopeptide ligase (E3), ubiquitin protein ligase E3C, ubiquitin-protein ligase E3C | |
| Between 725 and 775 | |
| 100 μg | |
| Cell Biology | |
| 9690 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction