missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE3C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20790
This item is not returnable.
View return policy
Description
UBE3C Polyclonal specifically detects UBE3C in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| UBE3C | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EC 6.3.2.-, KIAA0010KIAA10ubiquitin-protein isopeptide ligase (E3), ubiquitin protein ligase E3C, ubiquitin-protein ligase E3C | |
| Rabbit | |
| 124 kDa | |
| 0.1 mL | |
| Primary | |
| Species reactivity based on sequence identity: Chicken (87%), Bovine (89%), Xenopus tropicalis (80%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q15386 | |
| UBE3C | |
| Recombinant protein encompassing a sequence within the center region of human UBE3C. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9690 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction