missing translation for 'onlineSavingsMsg'
Learn More

UBE3B Antibody (OTI3F10) [Alexa Fluor™ 405], Novus Biologicals™

Product Code. 30689483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689483 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689483 Supplier Novus Biologicals Supplier No. NBP344687AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE3B Monoclonal antibody specifically detects UBE3B in Human,Mouse,Rat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE3B
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3F10
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp586K2123, DKFZp686A1051, EC 6.3.2, EC 6.3.2.-, FLJ45294, MGC131858, MGC78388, ubiquitin protein ligase E3B, ubiquitin-protein ligase E3B
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 768-1068 of human UBE3B (NP_569733) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89910
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.