missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2Z Monoclonal Antibody (K52034_14G2), Invitrogen™

Product Code. 30207994
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207994 50 μL 50µg
30196508 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30207994 Supplier Invitrogen™ Supplier No. MA555904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: WDGERTAPQC LLRIKRDIMS IYKEPPPGMF VVPDTVDMTK IHALITGPFD TPYEGGFFLF VFRCPPDYPI HPPRVKLMTT GNNTVRFNPN FYRNGKVCLS ILGTWTGPAW SPAQSISSVL ISIQSLMTEN PYHNEPGFEQ ERHPGDSKNY NECIRHETIR VAVCDMMEGK CPCPEPLRGV MEKSFLEYYD FYEVACKDRL HLQGQTMQDP FGEKRGHFDY QSLLMRLGLI RQKVLERLHN ENAEMDSDSS

This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2Z
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K52034_14G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene UBE2Z
Gene Accession No. Q9H832
Gene Alias AW049199; C030047H17Rik; D11Moh35; E2 ubiquitin-conjugating enzyme Z; HOYS7; RGD1308347; uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; UBE2Z; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2 Z; ubiquitin conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); ubiquitin-protein ligase Z; Use1
Gene Symbols UBE2Z
Host Species Mouse
Immunogen Recombinant human UBE2Z (immunogen range is 91-340).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65264
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.