missing translation for 'onlineSavingsMsg'
Learn More

UBE2W Antibody (7G4), Novus Biologicals™

Product Code. 18381969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381969 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381969 Supplier Novus Biologicals Supplier No. H00055284M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2W Monoclonal antibody specifically detects UBE2W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2W
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 7G4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001001481
Gene Alias EC 6.3.2.19, FLJ11011, hUBC-16, probable ubiquitin-conjugating enzyme E2 W, Ubiquitin carrier protein W, ubiquitin-conjugating enzyme 16, ubiquitin-conjugating enzyme E2W (putative), Ubiquitin-protein ligase W
Host Species Mouse
Immunogen UBE2W (NP_001001481, 17 a.a. ~ 112 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KRLQKELLALQNDPPPGMTLNEKSVQNSITQWIVDMEGAPGTLYEGEKFQLLFKFSSRYPFDSPQVMFTGENIPVHPHVYSNGHICLSILTEDWSP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 55284
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.