missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBE2W Monoclonal antibody specifically detects UBE2W in Human samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | mFluor Violet 500 SE |
| Gene Alias | EC 6.3.2.19, FLJ11011, hUBC-16, probable ubiquitin-conjugating enzyme E2 W, Ubiquitin carrier protein W, ubiquitin-conjugating enzyme 16, ubiquitin-conjugating enzyme E2W (putative), Ubiquitin-protein ligase W |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?