missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ UBE2V1 Recombinant Protein

Product Code. 16101712
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16101712

Brand: Abnova™ H00007335P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human UBE2V1 full-length ORF recombinant protein with GST-tag at N-terminal

Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein encoded by this gene is located in the nucleus and can cause transcriptional activation of the human FOS proto-oncogene. It is thought to be involved in the control of differentiation by altering cell cycle behavior. Multiple alternatively spliced transcripts encoding different isoforms have been described for this gene. A pseudogene has been identified which is also located on chromosome 20. Co-transcription of this gene and the neighboring upstream gene generates a rare transcript (Kua-UEV), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.

  • Theoretical MW (kDa): 41.91
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MAATTGSGVKVPRNFRLLEELEEGQKGVGDGTVSWGLEDDEDMTLTRWTGMIIGPPRTIYENRIYSLKIECGPKYPEAPPFVRFVTKINMNGVNSSNGVVDPRAISVLAKWQNSYSIKVVLQELRRLMMSKENMKLPQPPEGQCYSN

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH00468
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 7335
Molecular Weight (g/mol) 41.6
Name UBE2V1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MAATTGSGVKVPRNFRLLEELEEGQKGVGDGTVSWGLEDDEDMTLTRWTGMIIGPPRTIYENRIYSLKIECGPKYPEAPPFVRFVTKINMNGVNSSNGVVDPRAISVLAKWQNSYSIKVVLQELRRLMMSKENMKLPQPPEGQCYSN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CIR1/CROC-1/CROC1/UBE2V/UEV-1/UEV1/UEV1A
Common Name UBE2V1
Gene Symbol UBE2V1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.