missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2V1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565729
This item is not returnable.
View return policy
Description
Immunogen sequence: LLEELEEGQKG VGDGTVSWGL EDDEDMTLTR WTGMIIGPPR Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined.
Specifications
| UBE2V1 | |
| Polyclonal | |
| Unconjugated | |
| Ube2v1 | |
| 0610011J09Rik; AI256840; CIR1; CROC1; CROC-1; CROC1B; CROC-1B; CROC-1UBE2V; D7Bwg1382e; DJ1185N5.1.3; DNA-binding protein; KUA-UEV; P/OKcl.19; TRAF6-regulated IKK activator 1 beta Uev1A; UBE2V; Ube2v1; ubiquitin conjugating enzyme E2 V1; ubiquitin conjugating enzyme E2 variant 1; ubiquitin-conjugating enzyme E2 variant 1; UEV1; UEV-1; UEV1A; UEV-1CIR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7335 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13404 | |
| Ube2v1 | |
| Recombinant Human UBE2V1. Recombinant protein control fragment (Product #RP-104949). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction