missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2V1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828292
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828292 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828292 Supplier Invitrogen™ Supplier No. PA520081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Jurkat cell lysate. PA5-20081 can be used with blocking peptide PEP-0199.

The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2V1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ube2v1
Gene Accession No. Q13404, Q9CZY3
Gene Alias 0610011J09Rik; AI256840; CIR1; CROC1; CROC-1; CROC1B; CROC-1B; CROC-1UBE2V; D7Bwg1382e; DJ1185N5.1.3; DNA-binding protein; KUA-UEV; P/OKcl.19; TRAF6-regulated IKK activator 1 beta Uev1A; UBE2V; Ube2v1; ubiquitin conjugating enzyme E2 V1; ubiquitin conjugating enzyme E2 variant 1; ubiquitin-conjugating enzyme E2 variant 1; UEV1; UEV-1; UEV1A; UEV-1CIR1
Gene Symbols Ube2v1
Host Species Rabbit
Immunogen A peptide corresponding to 15 amino acids near the C-terminus of human UEV1A.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296390, 66589, 7335
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.