missing translation for 'onlineSavingsMsg'
Learn More

UBE2T Antibody (OTI2F5), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18433286 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433286 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433286 Supplier Novus Biologicals Supplier No. NBP274750AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2T Monoclonal antibody specifically detects UBE2T in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2T
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI2F5
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2T (NP_054895) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29089
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.