missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2T Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38343-100ul
This item is not returnable.
View return policy
Description
UBE2T Polyclonal antibody specifically detects UBE2T in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| UBE2T | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human UBE2T (NP_054895.1).,, Sequence:, MQRASRLKRELHMLATEPPPGITCWQDKDQMDDLRAQILGGANTPYEKGVFKLEVIIPERYPFEPPQIRFLTPIYHPNIDSAGRICLDVLKLPPKGAWRPSLNIATVLTSIQLLMSEPNPDDPLMADISSEFKYNKPAFLKNARQWTEKHARQKQKADEEEMLDNLPEAGDSRVHNSTQKRKASQLVGIEKKFHPDV | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29089 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction