missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBE2T Monoclonal antibody specifically detects UBE2T in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | UBE2T |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 4G1-4C2 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunoprecipitation |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH04152 |
| Gene Alias | Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?