missing translation for 'onlineSavingsMsg'
Learn More

UBE2S Antibody (2C12), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18204895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18204895 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18204895 Supplier Novus Biologicals Supplier No. NBP141126AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2S Monoclonal specifically detects UBE2S in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2S
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2C12
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q16763
Gene Alias E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S
Gene Symbols UBE2S
Host Species Mouse
Immunogen Recombinant human UBE2S (1-222aa) purified from E. coli (NP_055316.2).
Molecular Weight of Antigen 24 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27338
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.