missing translation for 'onlineSavingsMsg'
Learn More

UBE2R2 Antibody (5E7), Novus Biologicals™

Product Code. 18389309 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389309 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389309 Supplier Novus Biologicals Supplier No. H00054926M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2R2 Monoclonal antibody specifically detects UBE2R2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2R2
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 5E7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH47584
Gene Alias CDC34BEC 6.3.2.19, E2-CDC34B, FLJ20419, MGC10481, UBC3BUbiquitin-protein ligase R2, Ubiquitin carrier protein R2, ubiquitin-conjugating enzyme E2 R2, Ubiquitin-conjugating enzyme E2-CDC34B, ubiquitin-conjugating enzyme E2R 2, ubiquitin-conjugating enzyme UBC3B
Host Species Mouse
Immunogen UBE2R2 (AAH47584, 1 a.a. ~ 238 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAQQQMTSSQKALMLELKSLQEEPVEGFRITLVDESDLYNWEVAIFGPPNTLYEGGYFKAHIKFPIDYPYSPPTFRFLTKMWHPNIYENGDVCISILHPPVDDPQSGELPSERWNPTQNVRTILLSVISLLNEPNTFSPANVDASVMFRKWRDSKGKDKEYAEIIRKQVSATKAEAEKDGVKVPTTLAEYCIKTKVPSNDNSSDLLYDDLYDDDIDDEDEEEEDADCYDDDDSGNEES
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 54926
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.