missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2Q2 Antibody (2H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00092912-M04
This item is not returnable.
View return policy
Description
UBE2Q2 Monoclonal antibody specifically detects UBE2Q2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| UBE2Q2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp762C143, EC 6.3.2.19, LOC92912, Ubiquitin carrier protein Q2, ubiquitin-conjugating enzyme E2 Q2, ubiquitin-conjugating enzyme E2Q (putative) 2, ubiquitin-conjugating enzyme E2Q family member 2, Ubiquitin-protein ligase Q2 | |
| UBE2Q2 (NP_775740, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSVSGLKAELKFLASIFDKNHERFRIVSWKLDELHCQFLVPQQGSPHSLPPPLTLHCNITESYPSSSPIWFVDSEDPNLTSVLERLEDTKNNNLLRQQLKWLICELCSLY | |
| 0.1 mg | |
| Cell Biology | |
| 92912 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 2H3 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_775740 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction