missing translation for 'onlineSavingsMsg'
Learn More

UBE2O Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18380527 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380527 100 μL 100µL
18351756 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18380527 Supplier Novus Biologicals Supplier No. NBP303216100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2O Polyclonal specifically detects UBE2O in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2O
Applications Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias E2-230K, EC 6.3.2.19, FLJ12878, KIAA1734, Ubiquitin carrier protein O, ubiquitin-conjugating enzyme E2 O, Ubiquitin-conjugating enzyme E2 of 230 kDa, Ubiquitin-conjugating enzyme E2-230K, ubiquitin-conjugating enzyme E2O, Ubiquitin-protein ligase O
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1073-1292 of human UBE2O (NP_071349.3).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63893
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.