missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2N/Ubc13 Rabbit anti-Human, Mouse, Rat, Clone: 10O9Z5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16793-20UL
This item is not returnable.
View return policy
Description
UBE2N/Ubc13 Monoclonal antibody specifically detects UBE2N/Ubc13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| UBE2N/Ubc13 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 10O9Z5 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| bendless-like ubiquitin conjugating enzyme, Bendless-like ubiquitin-conjugating enzyme, BLU, EC 6.3.2.19, MGC131857, MGC8489, UBC13, UbcH-ben, Ubiquitin carrier protein N, ubiquitin-conjugating enzyme E2 N, ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13), ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast), Ubiquitin-protein ligase N, yeast UBC13 homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBE2N/Ubc13 (P61088). | |
| 20 μg | |
| Cancer | |
| 7334 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction