missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2M/Ubc12 Antibody (OTI2D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74748CL1
This item is not returnable.
View return policy
Description
UBE2M/Ubc12 Monoclonal antibody specifically detects UBE2M/Ubc12 in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| UBE2M/Ubc12 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
| Full length human recombinant protein of human UBE2M(NP_003960) produced in HEK293T cell. | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 9040 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction