missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2M/Ubc12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£180.00 - £248.00
Specifications
| Antigen | UBE2M/Ubc12 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698871
|
Novus Biologicals
NBP2-97606-100ul |
100 μg |
£248.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642932
|
Novus Biologicals
NBP2-97606-50ul |
50 μg |
£180.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE2M/Ubc12 Polyclonal antibody specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| UBE2M/Ubc12 | |
| Western Blot, Immunoprecipitation | |
| Purified | |
| RUO | |
| Human | |
| EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
| Produced in rabbits immunized with E. coli-derived Human UBE2M/Ubc12 fragment. | |
| Polyclonal | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins | |
| PBS (pH 7.0) | |
| 9040 | |
| IgG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title