missing translation for 'onlineSavingsMsg'
Learn More

UBE2M/Ubc12 Antibody, Biotin, Novus Biologicals™

Product Code. 18321042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321042 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321042 Supplier Novus Biologicals Supplier No. NBP300093B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2M/Ubc12 Polyclonal specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2M/Ubc12
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UBE2M/Ubc12 (P61081; Met1-Lys183).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 9040
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.