missing translation for 'onlineSavingsMsg'
Learn More

UBE2K/E2-25K Antibody (1D10), Novus Biologicals™

Product Code. 18374078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374078 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374078 Supplier Novus Biologicals Supplier No. H00003093M13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2K/E2-25K Monoclonal antibody specifically detects UBE2K/E2-25K in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2K/E2-25K
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1D10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. BC006419
Gene Alias E2(25K), E2-25K, EC 6.3.2.19, HIP-2, huntingtin interacting protein 2, Huntingtin-interacting protein 2, HYPG, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 K, Ubiquitin-conjugating enzyme E2(25K), Ubiquitin-conjugating enzyme E2-25 kDa, Ubiquitin-conjugating enzyme E2-25K, ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast), ubiquitin-protein ligase
Host Species Mouse
Immunogen UBE2K (-, 1 a.a. ~ 40 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLAMSPPAADSYPPPPPPLRPPPPPPPLPPPPPPSAIQT*
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3093
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.