missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2J1 Antibody (OTI3D1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74747CL1
This item is not returnable.
View return policy
Description
UBE2J1 Monoclonal antibody specifically detects UBE2J1 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| UBE2J1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CGI-76, EC 6.3.2.19, HSPC153, HSPC205, HSU93243, HsUBC6e, MGC12555, NCUBE-1, NCUBE1HSUBC6e, Non-canonical ubiquitin-conjugating enzyme 1, non-canonical ubquitin conjugating enzyme 1, Ubc6p, ubiquitin-conjugating enzyme E2 J1, ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast), Yeast ubiquitin-conjugating enzyme UBC6 homolog E | |
| Full length human recombinant protein of human UBE2J1(NP_057105) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI3D1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 51465 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction