missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15685-1-AP-150UL
This item is not returnable.
View return policy
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein sequence is 100% identical to the mouse homolog and 98% identical to the frog and zebrafish homologs. Three alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.Specifications
| UBE2H | |
| Polyclonal | |
| Unconjugated | |
| UBE2H | |
| E2 20K, UBC8, UBCH, UBCH2, UBE2H, Ubiquitin carrier protein H, Ubiquitin protein ligase H | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22214, 296956, 7328 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62256, P62257 | |
| UBE2H | |
| UBE2H Fusion Protein Ag8187 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction