missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2G1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12012-1-AP-20UL
This item is not returnable.
View return policy
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2G1 is a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 98-100% sequence identity with the zebrafish, frog, rat and mouse counterparts, which indicates that this enzyme is highly conserved in eukaryotes.Specifications
| UBE2G1 | |
| Polyclonal | |
| Unconjugated | |
| UBE2G1 | |
| E217K, UBC7, UBE2G, UBE2G1, Ubiquitin carrier protein G1, Ubiquitin protein ligase G1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64631, 67128, 7326 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62253, P62254, P62255 | |
| UBE2G1 | |
| UBE2G1 Fusion Protein Ag2625 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction