missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2G1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116099
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total UBE2G1.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2G1 is a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 98-100% sequence identity with the zebrafish, frog, rat and mouse counterparts, which indicates that this enzyme is highly conserved in eukaryotes.
Specifications
| UBE2G1 | |
| Polyclonal | |
| Unconjugated | |
| UBE2G1 | |
| 2700059C12Rik; AI256795; AU014992; AW552068; D130023C12Rik; E2 ubiquitin-conjugating enzyme G1; E217K; Ubc7; UBE2G; UBE2G1; ubiqu; Ubiquitin carrier protein G1; ubiquitin conjugating enzyme E2 G1; ubiquitin conjugating enzyme E2G 1; ubiquitin-conjugating enzyme E2 G1; Ubiquitin-conjugating enzyme E2 G1, N-terminally processed; ubiquitin-conjugating enzyme E2G 1; ubiquitin-conjugating enzyme E2G 1 (homologous to C. elegans UBC7); ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, C. elegans); ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast); ubiquitin-conjugating enzyme UBC7; ubiquitin-protein ligase G1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64631, 67128, 7326 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P62253, P62254, P62255 | |
| UBE2G1 | |
| A synthesized peptide derived from human UBE2G1(Accession P62253), corresponding to amino acid residues I75-I125. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction