missing translation for 'onlineSavingsMsg'
Learn More

UBE2G1 Antibody (1C12-1B2), Novus Biologicals™

Product Code. 18397697 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397697 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397697 Supplier Novus Biologicals Supplier No. H00007326M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2G1 Monoclonal antibody specifically detects UBE2G1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2G1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1C12-1B2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH02775
Gene Alias EC 6.3.2.19, UBC7, UBE2GE217K, Ubiquitin carrier protein G1, ubiquitin-conjugating enzyme E2 G1, ubiquitin-conjugating enzyme E2G 1 (homologous to C. elegans UBC7), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, C. elegans), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast), Ubiquitin-protein ligase G1
Host Species Mouse
Immunogen UBE2G1 (AAH02775, 1 a.a. ~ 170 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTELQSALLLRRQLAELNKNPVEGFSAGLIDDNDLYRWEVLIIGPPDTLYEGGVFKAHLTFPKDYPLRPPKMKFITEIWHPNVDKNGDVCISILHEPGEDKYGYEKPEERWLPIHTVETIMISVISMLADPNGDSPANVDAAKEWREDRNGEFKRKVARCVRKSQETAFE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7326
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.