missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UBE2F/NCE2 Polyclonal antibody specifically detects UBE2F/NCE2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Classification | Polyclonal |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 6.3.2, EC 6.3.2.-, MGC18120, NCE2Ubiquitin-conjugating enzyme E2 F, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme, NEDD8 protein ligase UBE2F, NEDD8-conjugating enzyme 2, NEDD8-conjugating enzyme UBE2F, ubiquitin-conjugating enzyme E2F (putative) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?