missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2E2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548639
This item is not returnable.
View return policy
Description
ubiquitin-conjugating enzyme E2 (UBE2E2) is a ubiquitin conjugating enzyme.
Specifications
| UBE2E2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q91W82, Q96LR5 | |
| UBE2E2 | |
| KLH conjugated synthetic peptide between 16-43 amino acids from the N-terminal region of human UBE2E2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| UBE2E2 | |
| BC016265; E2 ubiquitin-conjugating enzyme E2; FLJ25157; TBC1 domain family, member 12; UB2E2; UBCH8; UBE2E2; Ubiquitin carrier protein E2; ubiquitin conjugating enzyme E2 E2; ubiquitin conjugating enzyme E2E 2; ubiquitin-conjugating enzyme E2 E2; ubiquitin-conjugating enzyme E2E 2; ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast); ubiquitin-protein ligase E2; wu:fb38h07; zgc:92467 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 218793, 7325 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction