Learn More
UBE2E1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55457-1-AP-150UL
This item is not returnable.
View return policy
Description
The UBE2E1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human UBE2E1. This antibody recognizes human antigen. The UBE2E1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| UBE2E1 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| 150 μL | |
| 7324 | |
| IgG |