missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2D4/Ubch5d Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00095-50ul
This item is not returnable.
View return policy
Description
UBE2D4/Ubch5d Polyclonal antibody specifically detects UBE2D4/Ubch5d in Human samples. It is validated for ELISA
Specifications
| UBE2D4/Ubch5d | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| EC 6.3.2.19, FLJ32004, HBUCE1Ubiquitin carrier protein D4, UBCH5D, ubiquitin-conjugating enzyme E2 D4, ubiquitin-conjugating enzyme E2D 4 (putative), ubiquitin-conjugating enzyme HBUCE1, Ubiquitin-protein ligase D4 | |
| Produced in rabbits immunized with purified, recombinant Human UBE2D4/Ubch5d (Uniprot#: Q9Y2X8; Met1-Met1474) | |
| 50 μg | |
| Cell Biology | |
| 51619 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction