missing translation for 'onlineSavingsMsg'
Learn More

UBE2D4/Ubch5d Antibody (129), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18394441 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394441 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394441 Supplier Novus Biologicals Supplier No. NBP290194AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

UBE2D4/Ubch5d Monoclonal specifically detects UBE2D4/Ubch5d in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D4/Ubch5d
Applications ELISA
Classification Monoclonal
Clone 129
Conjugate Alexa Fluor 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, FLJ32004, HBUCE1Ubiquitin carrier protein D4, UBCH5D, ubiquitin-conjugating enzyme E2 D4, ubiquitin-conjugating enzyme E2D 4 (putative), ubiquitin-conjugating enzyme HBUCE1, Ubiquitin-protein ligase D4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human UBE2D4/Ubch5d (Q9Y2X8; Met1-Met1474).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51619
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.