missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2D4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15475-1-AP-150UL
This item is not returnable.
View return policy
Description
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro able to promote polyubiquitination using all 7 ubiquitin Lys residues, but may prefer ′Lys-11′ and ′Lys-48′-linked polyubiquitination.Specifications
| UBE2D4 | |
| Polyclonal | |
| Unconjugated | |
| UBE2D4 | |
| HBUCE1, UBCH5D, UBE2D4, Ubiquitin carrier protein D4, Ubiquitin protein ligase D4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51619, 546638, 79435 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y2X8 | |
| UBE2D4 | |
| UBE2D4 Fusion Protein Ag7779 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction