missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2C Recombinant Rabbit Monoclonal Antibody (SAIC-41C-6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551837
This item is not returnable.
View return policy
Description
Recognises human UBE2C.
This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is required for the destruction of mitotic cyclins and for cell cycle progression, and may be involved in cancer progression. Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-11'- and 'Lys-48'-linked polyubiquitination. Acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated ubiquitin ligase that controls progression through mitosis. Acts by initiating 'Lys-11'-linked polyubiquitin chains on APC/C substrates, leading to the degradation of APC/C substrates by the proteasome and promoting mitotic exit.
Specifications
| UBE2C | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O00762 | |
| UBE2C | |
| Peptide LSLEFPSGYPYNAPTVK derived from the Ubiquitin-conjugating enzyme E2C, conjugated to KLH (aa 81-97). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
| SAIC-41C-6 | |
| Unconjugated | |
| UBE2C | |
| (E3-independent) E2 ubiquitin-conjugating enzyme C; 1110015A16Rik; cyclin-selective ubiquitin carrier protein; D2Ertd695e; dJ447F3.2; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; RP3-447F3.1; ubcH10; UBE2C; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2 C; ubiquitin conjugating enzyme E2C; ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; ubiquitin-protein ligase C | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11065 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction