missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH8/Ube2L6 Antibody (k1H3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04351F
This item is not returnable.
View return policy
Description
UbcH8/Ube2L6 Monoclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| UbcH8/Ube2L6 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| UBE2L6 | |
| Recombinant human UBE2L6 (1-152aa) purified from E. coli | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9246 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| k1H3 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human UBE2L6 mAb, clone k1H3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human UBE2L6 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction