missing translation for 'onlineSavingsMsg'
Learn More

UbcH8/Ube2L6 Antibody, HRP, Novus Biologicals™

Product Code. 18391552 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391552 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391552 Supplier Novus Biologicals Supplier No. NBP300032H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UbcH8/Ube2L6 Polyclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH8/Ube2L6
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UbcH8/Ube2L6 (O14933-1; Met1-Ser153).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9246
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.