missing translation for 'onlineSavingsMsg'
Learn More

UbcH8/Ube2L6 Antibody (140), HRP, Novus Biologicals™

Product Code. 18389650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389650 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389650 Supplier Novus Biologicals Supplier No. NBP290137H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

UbcH8/Ube2L6 Monoclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH8/Ube2L6
Applications ELISA
Classification Monoclonal
Clone 140
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human UbcH8/Ube2L6 (O14933-1; Met1-Ser153).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9246
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.