missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH7/UBE2L3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99646-100ul
This item is not returnable.
View return policy
Description
UbcH7/UBE2L3 Polyclonal antibody specifically detects UbcH7/UBE2L3 in Human samples. It is validated for Western Blot
Specifications
| UbcH7/UBE2L3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| E2-F1, EC 6.3.2.19, L-UBC, UBCE7, UbcH7, UBCH7UbcM4, Ubiquitin carrier protein L3, ubiquitin-conjugating enzyme E2 L3, Ubiquitin-conjugating enzyme E2-F1, ubiquitin-conjugating enzyme E2L 3, ubiquitin-conjugating enzyme UBCH7, Ubiquitin-protein ligase L3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human UbcH7/UBE2L3. | |
| 100 μg | |
| Biologically Active Proteins, Cancer, DNA Repair, Tumor Suppressors | |
| 7332 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction