missing translation for 'onlineSavingsMsg'
Learn More

UbcH6/UBE2E1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18416997 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416997 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416997 Supplier Novus Biologicals Supplier No. NBP178983DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UbcH6/UBE2E1 Polyclonal antibody specifically detects UbcH6/UBE2E1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH6/UBE2E1
Applications Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot (Negative)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, UbcH6, Ubiquitin carrier protein E1, ubiquitin-conjugating enzyme E2 E1, ubiquitin-conjugating enzyme E2E 1 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E1
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human UBCH6 protein (between residues 1-50) [UniProt P51965]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7324
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.