missing translation for 'onlineSavingsMsg'
Learn More

UbcH6/UBE2E1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18390451 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390451 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390451 Supplier Novus Biologicals Supplier No. NBP178983AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UbcH6/UBE2E1 Polyclonal specifically detects UbcH6/UBE2E1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH6/UBE2E1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 6.3.2.19, UbcH6, Ubiquitin carrier protein E1, ubiquitin-conjugating enzyme E2 E1, ubiquitin-conjugating enzyme E2E 1 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E1
Gene Symbols UBE2E1
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human UBCH6 protein (between residues 1-50) [UniProt P51965]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7324.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.