missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ UbcH5c/UBE2D3 Antibody (S2), Novus Biologicals™

Product Code. 18344788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344788 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344788 Supplier Novus Biologicals™ Supplier No. H00007323M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UbcH5c/UBE2D3 Monoclonal antibody specifically detects UbcH5c/UBE2D3 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH5c/UBE2D3
Applications ELISA, Western Blot
Classification Monoclonal
Clone S2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH03395
Gene Alias E2(17)KB 3, E2(17)KB3, EC 6.3.2.19, MGC43926, MGC5416, UBC4/5, UBC5C, UBCH5C, Ubiquitin carrier protein D3, ubiquitin-conjugating enzyme E2 D3, Ubiquitin-conjugating enzyme E2(17)KB 3, Ubiquitin-conjugating enzyme E2-17 kDa 3, ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D3
Host Species Mouse
Immunogen UBE2D3 (AAH03395, 1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MALKRINKELSDLARDPPAQCSAGPVGDDMFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTRIYHPNINSNGSICLDILRSQWSPALTISKVLLSICSLLCDPNPDDPLVPEIARIYKTDRDKYNRISREWTQKYAM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7323
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.