missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH5c/UBE2D3 Antibody (4C1-1E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00007323-M01
This item is not returnable.
View return policy
Description
UbcH5c/UBE2D3 Monoclonal antibody specifically detects UbcH5c/UBE2D3 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| UbcH5c/UBE2D3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| E2(17)KB 3, E2(17)KB3, EC 6.3.2.19, MGC43926, MGC5416, UBC4/5, UBC5C, UBCH5C, Ubiquitin carrier protein D3, ubiquitin-conjugating enzyme E2 D3, Ubiquitin-conjugating enzyme E2(17)KB 3, Ubiquitin-conjugating enzyme E2-17 kDa 3, ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D3 | |
| UBE2D3 (AAH03395, 1 a.a. ~ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MALKRINKELSDLARDPPAQCSAGPVGDDMFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTRIYHPNINSNGSICLDILRSQWSPALTISKVLLSICSLLCDPNPDDPLVPEIARIYKTDRDKYNRISREWTQKYAM | |
| 0.1 mg | |
| Biologically Active Proteins | |
| 7323 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 4C1-1E3 | |
| Western Blot, ELISA, Sandwich ELISA | |
| AAH03395 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction