missing translation for 'onlineSavingsMsg'
Learn More

UbcH5a/UBE2D1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18380623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380623 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380623 Supplier Novus Biologicals Supplier No. NBP299796AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UbcH5a/UBE2D1 Polyclonal specifically detects UbcH5a/UBE2D1 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH5a/UBE2D1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UbcH5a/UBE2D1 (P51668; Ala2-Met147).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7321
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.