missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UbcH2/UBE2H Polyclonal antibody specifically detects UbcH2/UBE2H in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Applications | ELISA |
| Conjugate | Janelia Fluor 646 |
| Host Species | Rabbit |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?