missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH2/UBE2H Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93052-0.1ml
This item is not returnable.
View return policy
Description
UbcH2/UBE2H Polyclonal antibody specifically detects UbcH2/UBE2H in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UbcH2/UBE2H | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| EC 6.3.2.19, UBC8E2-20K, UBCH, UBCH2, Ubiquitin carrier protein H, ubiquitin-conjugating enzyme E2 H, Ubiquitin-conjugating enzyme E2-20K, ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8), ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast), Ubiquitin-protein ligase H | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-183 of human UBE2H (NP_003335.1). MSSPSPGKRRMDTDVVKLIESKHEVTILGGLNEFVVKFYGPQGTPYEGGVWKVRVDLPDKYPFKSPSIGFMNKIFHPNIDEASGTVCLDVINQTWTALYDLTNIFESFLPQLLAYPNPIDPLNGDAAAMYLHRPEEYKQKIKEYIQKYATEEALKEQEEGTGDSSSESSMSDFSEDEAQDMEL | |
| 0.1 mL | |
| Cell Biology | |
| 7328 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction